1-[(E)-2-(Furan-2-yl)ethenyl]-3-methylurea, tech
Lieferant: Apollo Scientific
OR32635-1G
APOSOR32635-1GEA
1570
EUR
APOSOR32635-1G
1-[(E)-2-(Furan-2-yl)ethenyl]-3-methylurea, tech
1-[(E)-2-(Furan-2-yl)ethenyl]-3-methylurea
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00794351 CAS-Nummer: 478030-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...