1-(2,6-Dimethylphenyl)-3-[(E)-2-(furan-2-yl)ethenyl]urea, tech

Lieferant: Apollo Scientific

OR32636-1G
APOSOR32636-1GEA 1570 EUR
APOSOR32636-1G
1-(2,6-Dimethylphenyl)-3-[(E)-2-(furan-2-yl)ethenyl]urea, tech
1-(2,6-Dimethylphenyl)-3-[(E)-2-(furan-2-yl)ethenyl]urea
Formel: C₁₅H₁₆N₂O₂
Molecular Weight: 256,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794359
CAS-Nummer: 478031-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR