[2-(Phenylsulphanyl)pyridin-3-yl]methanol, tech
Lieferant: Apollo Scientific
OR32638-1G
APOSOR32638-1GEA
1570
EUR
APOSOR32638-1G
[2-(Phenylsulphanyl)pyridin-3-yl]methanol, tech
[2-(Phenylsulphanyl)pyridin-3-yl]methanol
Formel:
C₁₂H₁₁NOS Molecular Weight: 217,287 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00794865 CAS-Nummer: 43003-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...