4-(Benzenesulphonyl)-3-methyl-3,4-dihydro-2H-1,4-benzoxazine, tech

Lieferant: Apollo Scientific

OR32641-1G
APOSOR32641-1GEA 1570 EUR
APOSOR32641-1G
4-(Benzenesulphonyl)-3-methyl-3,4-dihydro-2H-1,4-benzoxazine, tech
4-(Benzenesulphonyl)-3-methyl-3,4-dihydro-2H-1,4-benzoxazine
Formel: C₁₅H₁₅NO₃S
Molecular Weight: 289.35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02083423
CAS-Nummer: 478031-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR