8-Amino-3-(4-chlorphenyl)imidazo[1,2-a]pyridin 95%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Chlorophenyl)imidazo[1,2-a]pyridin-8-amine

OR32642-1G
APOSOR32642-1GEA 488 EUR
APOSOR32642-1G
8-Amino-3-(4-chlorphenyl)imidazo[1,2-a]pyridin 95%
8-Amino-3-(4-chlorphenyl)imidazo[1,2-a]pyridin
Formel: C₁₃H₁₀ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00975015
CAS-Nummer: 338415-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR