2-Methyl-1-(4-methylbenzyl)-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 95%

Lieferant: Apollo Scientific

OR32643-1G
APOSOR32643-1GEA 427 EUR
APOSOR32643-1G
2-Methyl-1-(4-methylbenzyl)-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 95%
2-Methyl-1-(4-methylbenzyl)-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile
Formel: C₁₅H₁₆N₂O
Molecular Weight: 240,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975017
CAS-Nummer: 338415-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR