5-Acetyl-6-[2-(dimethylamino)vinyl]-2(1H)-pyridinone 95%
Lieferant: Apollo Scientific
Synonyme:
5-Acetyl-6-[(E)-2-(dimethylamino)ethenyl]-1,2-dihydropyridin-2-one
OR32645-1G
APOSOR32645-1GEA
1070
EUR
APOSOR32645-1G
5-Acetyl-6-[2-(dimethylamino)vinyl]-2(1H)-pyridinone 95%
5-Acetyl-6-[2-(dimethylamino)vinyl]-2(1H)-pyridinone
Formel:
C₁₁H₁₄N₂O₂ Molecular Weight: 206,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01814813 CAS-Nummer: 88877-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...