5-Methyl[1,6]naphthyridin-2(1H)-one 95%
Lieferant: Apollo Scientific
Synonyme:
5-Methyl-1,2-dihydro-1,6-naphthyridin-2-one
OR32646-1G
APOSOR32646-1GEA
671
EUR
APOSOR32646-1G
5-Methyl[1,6]naphthyridin-2(1H)-one 95%
5-Methyl[1,6]naphthyridin-2(1H)-one
Formel:
C₉H₈N₂O Molecular Weight: 160,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00867139 CAS-Nummer: 88296-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...