Ethyl 2-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate 95%

Lieferant: Apollo Scientific

OR32648-1G
APOSOR32648-1GEA 94.4 EUR
APOSOR32648-1G
Ethyl 2-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate 95%
Ethyl 2-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate
Formel: C₉H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01871481
CAS-Nummer: 3424-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR