(2E)-3-(4-Methoxyphenyl)-1-(pyridin-2-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32649-1G
APOSOR32649-1GEA 1570 EUR
APOSOR32649-1G
(2E)-3-(4-Methoxyphenyl)-1-(pyridin-2-yl)prop-2-en-1-one, tech
(2E)-3-(4-Methoxyphenyl)-1-(pyridin-2-yl)prop-2-en-1-one
Formel: C₁₅H₁₃NO₂
Molecular Weight: 239,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315417
CAS-Nummer: 18451-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR