2-[2-(4-Methoxyphenyl)cyclopropanecarbonyl]pyridine, tech
Lieferant: Apollo Scientific
OR32651-1G
APOSOR32651-1GEA
1570
EUR
APOSOR32651-1G
2-[2-(4-Methoxyphenyl)cyclopropanecarbonyl]pyridine, tech
2-[2-(4-Methoxyphenyl)cyclopropanecarbonyl]pyridine
Formel:
C₁₆H₁₅NO₂ Molecular Weight: 253,296 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01315531 CAS-Nummer: 338415-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...