1-(4-Chlorophenyl)pentane-1,4-dione, tech

Lieferant: Apollo Scientific

OR32652-1G
APOSOR32652-1GEA 1570 EUR
APOSOR32652-1G
1-(4-Chlorophenyl)pentane-1,4-dione, tech
1-(4-Chlorophenyl)pentane-1,4-dione
Formel: C₁₁H₁₁ClO₂
Molecular Weight: 210,657 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316205
CAS-Nummer: 53842-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR