1-(Cyclopropylmethyl)-4-hydroxy-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one, tech
Lieferant: Apollo Scientific
OR32659-1G
APOSOR32659-1GEA
1570
EUR
APOSOR32659-1G
1-(Cyclopropylmethyl)-4-hydroxy-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one, tech
1-(Cyclopropylmethyl)-4-hydroxy-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one
Formel:
C₁₈H₁₉NO₂ Molecular Weight: 281,349 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02082727 CAS-Nummer: 478033-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...