3-Bromo-2-furaldehyde 96%

Lieferant: Apollo Scientific
Achtung

OR3266-1G OR3266-5G
APOSOR3266-1GEA 58 EUR
APOSOR3266-1G APOSOR3266-5G
3-Bromo-2-furaldehyde 96%
3-Bromo-2-furaldehyde
Formel: C₅H₃BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07368224
CAS-Nummer: 14757-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR