2-(Phenylamino)-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione, tech

Lieferant: Apollo Scientific

OR32661-1G
APOSOR32661-1GEA 1570 EUR
APOSOR32661-1G
2-(Phenylamino)-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione, tech
2-(Phenylamino)-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione
Formel: C₁₄H₁₄N₂O₂
Molecular Weight: 242,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01717970
CAS-Nummer: 61152-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR