Ethyl (benzylcarbamoyl)formate, tech

Lieferant: Apollo Scientific

OR32664-1G
APOSOR32664-1GEA 1570 EUR
APOSOR32664-1G
Ethyl (benzylcarbamoyl)formate, tech
Ethyl (benzylcarbamoyl)formate
Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00196026
CAS-Nummer: 7142-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR