1-(2-Methyl-1H-indol-3-yl)-2-(piperidin-1-yl)ethane-1,2-dione, tech

Lieferant: Apollo Scientific

OR32665-1G
APOSOR32665-1GEA 1570 EUR
APOSOR32665-1G
1-(2-Methyl-1H-indol-3-yl)-2-(piperidin-1-yl)ethane-1,2-dione, tech
1-(2-Methyl-1H-indol-3-yl)-2-(piperidin-1-yl)ethane-1,2-dione
Formel: C₁₆H₁₈N₂O₂
Molecular Weight: 270,326 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00654545
CAS-Nummer: 330965-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR