6-(4-Chlorobenzenesulphonyl)-1-oxa-6-azaspiro[2.5]octane, tech

Lieferant: Apollo Scientific

OR32666-1G
APOSOR32666-1GEA 1570 EUR
APOSOR32666-1G
6-(4-Chlorobenzenesulphonyl)-1-oxa-6-azaspiro[2.5]octane, tech
6-(4-Chlorobenzenesulphonyl)-1-oxa-6-azaspiro[2.5]octane
Formel: C₁₂H₁₄ClNO₃S
Molecular Weight: 287,762 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125157
CAS-Nummer: 861212-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR