(2E)-3-(2,4-Dichlorophenyl)prop-2-enehydrazide, tech

Lieferant: Apollo Scientific

OR32667-1G
APOSOR32667-1GEA 1570 EUR
APOSOR32667-1G
(2E)-3-(2,4-Dichlorophenyl)prop-2-enehydrazide, tech
(2E)-3-(2,4-Dichlorophenyl)prop-2-enehydrazide
Formel: C₉H₈Cl₂N₂O
Molecular Weight: 231.079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125159
CAS-Nummer: 876729-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR