4-Methyl-N-{1H-pyrrolo[2,3-b]pyridin-1-yl}benzamide, tech

Lieferant: Apollo Scientific

OR32668-1G
APOSOR32668-1GEA 1570 EUR
APOSOR32668-1G
4-Methyl-N-{1H-pyrrolo[2,3-b]pyridin-1-yl}benzamide, tech
4-Methyl-N-{1H-pyrrolo[2,3-b]pyridin-1-yl}benzamide
Formel: C₁₅H₁₃N₃O
Molecular Weight: 251,283 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03102808
CAS-Nummer: 861212-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR