Cyanomethyl (2Z)-3-phenylprop-2-enoate, tech

Lieferant: Apollo Scientific

OR32669-1G
APOSOR32669-1GEA 1570 EUR
APOSOR32669-1G
Cyanomethyl (2Z)-3-phenylprop-2-enoate, tech
Cyanomethyl (2Z)-3-phenylprop-2-enoate
Formel: C₁₁H₉NO₂
Molecular Weight: 187,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125161
CAS-Nummer: 21955-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR