1-{2-Methylimidazo[1,2-a]pyridin-3-yl}-2-(morpholin-4-yl)ethan-1-one, tech
Lieferant: Apollo Scientific
OR32670-1G
APOSOR32670-1GEA
1570
EUR
APOSOR32670-1G
1-{2-Methylimidazo[1,2-a]pyridin-3-yl}-2-(morpholin-4-yl)ethan-1-one, tech
1-{2-Methylimidazo[1,2-a]pyridin-3-yl}-2-(morpholin-4-yl)ethan-1-one
Formel:
C₁₄H₁₇N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03012040 CAS-Nummer: 383148-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...