1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one, tech

Lieferant: Apollo Scientific

OR32671-1G
APOSOR32671-1GEA 1570 EUR
APOSOR32671-1G
1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one, tech
1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one
Formel: C₁₀H₉ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00973744
CAS-Nummer: 154877-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR