Methyl 4-(morpholin-4-yl)benzoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-morpholinobenzoate

OR32672-1G
APOSOR32672-1GEA 180 EUR
APOSOR32672-1G
Methyl 4-(morpholin-4-yl)benzoate 95%
Methyl 4-(morpholin-4-yl)benzoate
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221.252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012127
CAS-Nummer: 23676-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR