4-(1H-1,3-Benzimidazol-1-yl)benzenecarbaldehyde 95%
Lieferant: Apollo Scientific
OR32676-1G
APOSOR32676-1GEA
0
EUR
APOSOR32676-1G
4-(1H-1,3-Benzimidazol-1-yl)benzenecarbaldehyde 95%
4-(1H-1,3-Benzimidazol-1-yl)benzenecarbaldehyde
Formel:
C₁₄H₁₀N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425762 CAS-Nummer: 90514-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...