[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol 95%

Lieferant: Apollo Scientific

OR32677-1G OR32677-5G
APOSOR32677-1GEA 270 EUR
APOSOR32677-1G APOSOR32677-5G
[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol 95%
[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol
Formel: C₁₄H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03425763
CAS-Nummer: 451485-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR