Methyl 2-(4-hydroxyphenoxy)propanoate, tech

Lieferant: Apollo Scientific

OR32680-1G
APOSOR32680-1GEA 1570 EUR
APOSOR32680-1G
Methyl 2-(4-hydroxyphenoxy)propanoate, tech
Methyl 2-(4-hydroxyphenoxy)propanoate
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00204480
CAS-Nummer: 60075-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR