2-Phenylimidazo[1,2-a]pyridine-6-carbonitrile, tech

Lieferant: Apollo Scientific

OR32682-1G
APOSOR32682-1GEA 1570 EUR
APOSOR32682-1G
2-Phenylimidazo[1,2-a]pyridine-6-carbonitrile, tech
2-Phenylimidazo[1,2-a]pyridine-6-carbonitrile
Formel: C₁₄H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03787072
CAS-Nummer: 214958-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR