4-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]pyrimidin-2-amine, tech

Lieferant: Apollo Scientific

OR32683-1G
APOSOR32683-1GEA 1570 EUR
APOSOR32683-1G
4-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]pyrimidin-2-amine, tech
4-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]pyrimidin-2-amine
Formel: C₁₆H₁₆N₄
Molecular Weight: 264,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03848450
CAS-Nummer: 865658-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR