6-Chloro-2-(2-chloroethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
Lieferant: Apollo Scientific
OR32684-1G
APOSOR32684-1GEA
1570
EUR
APOSOR32684-1G
6-Chloro-2-(2-chloroethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
6-Chloro-2-(2-chloroethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one
Formel:
C₁₀H₉Cl₂NO₂ Molecular Weight: 246,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787075 CAS-Nummer: 865658-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...