Pyrrolo[1,2-a]quinolin-6-amine, tech

Lieferant: Apollo Scientific

OR32687-1G
APOSOR32687-1GEA 1570 EUR
APOSOR32687-1G
Pyrrolo[1,2-a]quinolin-6-amine, tech
Pyrrolo[1,2-a]quinolin-6-amine
Formel: C₁₂H₁₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03848484
CAS-Nummer: 865658-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR