4,6-Dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32690-1G
APOSOR32690-1GEA 1570 EUR
APOSOR32690-1G
4,6-Dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile, tech
4,6-Dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile
Formel: C₁₂H₁₅N₃O
Molecular Weight: 217.267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00124668
CAS-Nummer: 142591-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR