4-(4-Chlorophenyl)-3-methyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR32692-1G
APOSOR32692-1GEA 0 EUR
APOSOR32692-1G
4-(4-Chlorophenyl)-3-methyl-1H-pyrazole 95%
4-(4-Chlorophenyl)-3-methyl-1H-pyrazole
Formel: C₁₀H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00179386
CAS-Nummer: 667400-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR