2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-hexahydro-1H-pyrrolo[1,2-c]imidazole-1,3-dione, tech

Lieferant: Apollo Scientific

OR32693-1G
APOSOR32693-1GEA 1570 EUR
APOSOR32693-1G
2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-hexahydro-1H-pyrrolo[1,2-c]imidazole-1,3-dione, tech
2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-hexahydro-1H-pyrrolo[1,2-c]imidazole-1,3-dione
Formel: C₁₄H₁₄N₂O₄
Molecular Weight: 274,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26383446
CAS-Nummer: 1796902-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR