Methyl 4-[(6-chloropyridin-3-yl)methoxy]benzoate, tech

Lieferant: Apollo Scientific

OR32699-1G
APOSOR32699-1GEA 0 EUR
APOSOR32699-1G
Methyl 4-[(6-chloropyridin-3-yl)methoxy]benzoate, tech
Methyl 4-[(6-chloropyridin-3-yl)methoxy]benzoate
Formel: C₁₄H₁₂ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04125299
CAS-Nummer: 865660-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR