1-(3-Aminophenyl)pyrrolidin-2-one 95%

Lieferant: Apollo Scientific

OR32702-1G
APOSOR32702-1GEA 180 EUR
APOSOR32702-1G
1-(3-Aminophenyl)pyrrolidin-2-one 95%
1-(3-Aminophenyl)pyrrolidin-2-one
Formel: C₁₀H₁₂N₂O
Molecular Weight: 176.22 g/mol
Schmelzpunkt: 29…32 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00468694
CAS-Nummer: 31992-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR