3-[(4-Hydroxy-3-methoxy-5-methylphenyl)methyl]-2,3-dihydro-1H-indol-2-one, tech

Lieferant: Apollo Scientific

OR32704-1G
APOSOR32704-1GEA 1570 EUR
APOSOR32704-1G
3-[(4-Hydroxy-3-methoxy-5-methylphenyl)methyl]-2,3-dihydro-1H-indol-2-one, tech
3-[(4-Hydroxy-3-methoxy-5-methylphenyl)methyl]-2,3-dihydro-1H-indol-2-one
Formel: C₁₇H₁₇NO₃
Molecular Weight: 283,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08281853
CAS-Nummer: 882747-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR