4-Amino-2-[2-(pyridin-2-yl)ethyl]-2,3-dihydro-1H-isoindol-1-one, tech

Lieferant: Apollo Scientific

OR32705-1G
APOSOR32705-1GEA 1570 EUR
APOSOR32705-1G
4-Amino-2-[2-(pyridin-2-yl)ethyl]-2,3-dihydro-1H-isoindol-1-one, tech
4-Amino-2-[2-(pyridin-2-yl)ethyl]-2,3-dihydro-1H-isoindol-1-one
Formel: C₁₅H₁₅N₃O
Molecular Weight: 253,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07021409
CAS-Nummer: 900015-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR