Methyl-2-phenylimidazo[1,2-a]pyridin-6-carboxylat 95%

Lieferant: Apollo Scientific

OR32707-1G
APOSOR32707-1GEA 671 EUR
APOSOR32707-1G
Methyl-2-phenylimidazo[1,2-a]pyridin-6-carboxylat 95%
Methyl-2-phenylimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₁₅H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07698590
CAS-Nummer: 962-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR