4-Bromo-2-methoxy-6-[(1Z)-(methoxyimino)methyl]phenol, tech

Lieferant: Apollo Scientific

OR32709-1G
APOSOR32709-1GEA 1570 EUR
APOSOR32709-1G
4-Bromo-2-methoxy-6-[(1Z)-(methoxyimino)methyl]phenol, tech
4-Bromo-2-methoxy-6-[(1Z)-(methoxyimino)methyl]phenol
Formel: C₉H₁₀BrNO₃
Molecular Weight: 260,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138190
CAS-Nummer: 338416-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR