(3Z)-4-(Dimethylamino)-3-[(4-methylphenyl)sulphanyl]but-3-en-2-one, tech

Lieferant: Apollo Scientific

OR32712-1G
APOSOR32712-1GEA 1570 EUR
APOSOR32712-1G
(3Z)-4-(Dimethylamino)-3-[(4-methylphenyl)sulphanyl]but-3-en-2-one, tech
(3Z)-4-(Dimethylamino)-3-[(4-methylphenyl)sulphanyl]but-3-en-2-one
Formel: C₁₃H₁₇NOS
Molecular Weight: 235,345 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138926
CAS-Nummer: 1164559-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR