[1-(3,4-Dichlorophenyl)-1H-1,2,3-triazol-4-yl]methanol 95%

Lieferant: Apollo Scientific

OR32717-1G
APOSOR32717-1GEA 1330 EUR
APOSOR32717-1G
[1-(3,4-Dichlorophenyl)-1H-1,2,3-triazol-4-yl]methanol 95%
[1-(3,4-Dichlorophenyl)-1H-1,2,3-triazol-4-yl]methanol
Formel: C₉H₇Cl₂N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00140934
CAS-Nummer: 338419-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR