Ethyl-2,4-dimethyl-7-phenylpyrrolo[1,2-a]pyrimidine-8-carboxylate 95%

Lieferant: Apollo Scientific

OR32721-1G
APOSOR32721-1GEA 1410 EUR
APOSOR32721-1G
Ethyl-2,4-dimethyl-7-phenylpyrrolo[1,2-a]pyrimidine-8-carboxylate 95%
Ethyl-2,4-dimethyl-7-phenylpyrrolo[1,2-a]pyrimidine-8-carboxylate
Formel: C₁₈H₁₈N₂O₂
Molecular Weight: 294,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171951
CAS-Nummer: 338419-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR