Methyl-2H-chromene-3-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2H-chromene-3-carboxylate
OR32724-1G
APOSOR32724-1GEA
671
EUR
APOSOR32724-1G
Methyl-2H-chromene-3-carboxylate 95%
Methyl-2H-chromene-3-carboxylate
Formel:
C₁₁H₁₀O₃ Molecular Weight: 190,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202022 CAS-Nummer: 36044-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...