Ethyl 8-methoxy-2H-chromene-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32726-1G
APOSOR32726-1GEA 1570 EUR
APOSOR32726-1G
Ethyl 8-methoxy-2H-chromene-3-carboxylate, tech
Ethyl 8-methoxy-2H-chromene-3-carboxylate
Formel: C₁₃H₁₄O₄
Molecular Weight: 234,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316444
CAS-Nummer: 112904-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR