(Pyridin-3-yl)methyl 8-methoxy-2H-chromene-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32727-1G
APOSOR32727-1GEA 1570 EUR
APOSOR32727-1G
(Pyridin-3-yl)methyl 8-methoxy-2H-chromene-3-carboxylate, tech
(Pyridin-3-yl)methyl 8-methoxy-2H-chromene-3-carboxylate
Formel: C₁₇H₁₅NO₄
Molecular Weight: 297,305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202026
CAS-Nummer: 338419-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR