Methyl-6-chloro-2H-chromene-3-carboxylate 95%
Lieferant: Apollo Scientific
OR32730-1G
APOSOR32730-1GEA
538
EUR
APOSOR32730-1G
Methyl-6-chloro-2H-chromene-3-carboxylate 95%
Methyl-6-chloro-2H-chromene-3-carboxylate
Formel:
C₁₁H₉ClO₃ Molecular Weight: 224,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202038 CAS-Nummer: 68281-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...