N-(4,6-Dimethylpyridin-2-yl)-3-oxobutanamide, tech

Lieferant: Apollo Scientific

OR32731-1G
APOSOR32731-1GEA 1570 EUR
APOSOR32731-1G
N-(4,6-Dimethylpyridin-2-yl)-3-oxobutanamide, tech
N-(4,6-Dimethylpyridin-2-yl)-3-oxobutanamide
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231511
CAS-Nummer: 23600-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR