2-(3-Methoxyphenyl)-2-(morpholin-4-yl)acetonitrile 95%

Lieferant: Apollo Scientific

OR32733-1G
APOSOR32733-1GEA 1120 EUR
APOSOR32733-1G
2-(3-Methoxyphenyl)-2-(morpholin-4-yl)acetonitrile 95%
2-(3-Methoxyphenyl)-2-(morpholin-4-yl)acetonitrile
Formel: C₁₃H₁₆N₂O₂
Molecular Weight: 232,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172615
CAS-Nummer: 123567-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR