3,5-Dichloro-1H-pyrrole-2,4-dicarbaldehyde, tech

Lieferant: Apollo Scientific

OR32736-1G
APOSOR32736-1GEA 1570 EUR
APOSOR32736-1G
3,5-Dichloro-1H-pyrrole-2,4-dicarbaldehyde, tech
3,5-Dichloro-1H-pyrrole-2,4-dicarbaldehyde
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172616
CAS-Nummer: 152420-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR